Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004440.1 | NP_001004440.1 | 3 | 270 | ||
NM_182759.2 | NP_877436.1 | 3 | 270 | |||
XM_005270766.3 | XP_005270823.1 | 4 | 557 | |||
XM_006710582.3 | XP_006710645.1 | 4 | 557 | |||
GenBank mRNA | AY325116.1 | - | 3 | 270 | ||
AY325119.1 | - | 3 | 270 | |||
BC112268.1 | - | 3 | 264 | 8327736 | ||
BC117479.1 | - | 3 | 264 | 40126030 | ||
BC143949.1 | - | 3 | 264 | |||
BC144042.1 | - | 3 | 253 | |||
BC144043.1 | - | 3 | 264 | 9052563 | ||
BQ638101.1 | - | 3 | 418 |