Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010883.2 | NP_001010883.2 | 2 | 254 | ||
GenBank mRNA | BC127898.1 | - | 2 | 254 | 40132918 | |
BC140863.1 | - | 2 | 110 | 9021731 | ||
BC140865.1 | - | 2 | 110 | 9021733 | ||
BC150530.1 | - | 2 | 254 | 40132919 |