Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122962.1 | NP_001116434.1 | 2 | 526 | ||
XM_005260708.4 | XP_005260765.1 | 2 | 335 | |||
XM_011529221.2 | XP_011527523.1 | 2 | 1122 | |||
GenBank mRNA | AK095499.1 | - | 2 | 335 | ||
BM285394.1 | - | 2 | 528 | |||
BQ635335.1 | - | 2 | 595 | |||
DC313096.1 | - | 2 | 424 | |||
DC358609.1 | - | 2 | 389 |