Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316745.1 | NP_001303674.1 | 7 | 1060 | ||
NM_173648.3 | NP_775919.3 | 7 | 1060 | |||
XM_011510991.2 | XP_011509293.1 | 7 | 1135 | |||
XM_011510992.2 | XP_011509294.1 | 7 | 1135 | |||
XM_011510993.2 | XP_011509295.1 | 7 | 1135 | |||
XM_017003867.1 | XP_016859356.1 | 7 | 1135 | |||
XM_017003868.1 | XP_016859357.1 | 7 | 1134 | |||
XM_017003869.1 | XP_016859358.1 | 7 | 1133 | |||
GenBank mRNA | AL832777.1 | - | 7 | 1060 |