Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289947.1 | NP_001276876.1 | 2 | 93 | ||
NM_001290030.1 | NP_001276959.1 | 7 | 880 | |||
XM_011510998.2 | XP_011509300.1 | 9 | 1012 | |||
XM_011510999.2 | XP_011509301.1 | 7 | 583 | |||
XM_011511001.2 | XP_011509303.1 | 7 | 1358 | |||
XM_017003875.1 | XP_016859364.1 | 8 | 1381 | |||
XM_017003876.1 | XP_016859365.1 | 7 | 396 | |||
XM_017003877.1 | XP_016859366.1 | 8 | 679 | |||
GenBank mRNA | BC127676.1 | - | 2 | 93 | 40127368 | |
BC150546.1 | - | 2 | 93 | |||
BQ429225.1 | - | 91 |