Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC66 SIRNA Assays ›
Gene Symbol:
CCDC66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
-
Chromosome Location:
Chr.3: 56557125 - 56621837 on Build GRCh38
UniGene ID:
Hs.476399
Species:
Human
siRNA ID
HSS154843
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012506.4
NP_001012524.4
16
2561
NM_001141947.1
NP_001135419.1
16
2580
NR_024460.1
16
2561
XM_005265081.3
XP_005265138.1
16
2598
XM_005265082.3
XP_005265139.1
16
2577
XM_005265083.3
XP_005265140.1
16
2469
XM_005265084.3
XP_005265141.1
16
2592
XM_011533614.2
XP_011531916.1
15
2603
XM_011533615.2
XP_011531917.1
15
2613
XM_011533616.2
XP_011531918.1
16
2537
XM_011533617.2
XP_011531919.1
17
2695
XM_011533619.2
XP_011531921.1
17
2598
XM_017006229.1
XP_016861718.1
16
2559
XM_017006230.1
XP_016861719.1
15
2581
XM_017006231.1
XP_016861720.1
15
2584
XM_017006232.1
XP_016861721.1
16
2451
XM_017006233.1
XP_016861722.1
16
2430
XM_017006234.1
XP_016861723.1
16
2619
XM_017006235.1
XP_016861724.1
17
2677
XM_017006236.1
XP_016861725.1
18
2690
XM_017006237.1
XP_016861726.1
18
2727
XM_017006238.1
XP_016861727.1
19
2868
XM_017006239.1
XP_016861728.1
13
1928
XM_017006240.1
XP_016861729.1
17
2631
XM_017006241.1
XP_016861730.1
18
2590
XM_017006242.1
XP_016861731.1
17
2453
XM_017006243.1
XP_016861732.1
16
2546
XM_017006244.1
XP_016861733.1
17
2682
XM_017006245.1
XP_016861734.1
15
2385
GenBank mRNA
AK096754.1
-
3
1896
AL832556.1
-
1
2285
AL832692.1
-
16
2501
AL833326.1
-
17
2598
BC047509.1
-
16
2529
5276957
BC132827.1
-
16
2550
40146849
BU674751.1
-
3
569
Back To Top
More Information
Gene Ontology Categories:
Function(s)
retinal rod cell development
detection of light stimulus involved in visual perception
post-embryonic retina morphogenesis in camera-type eye
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline