Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164674.1 | NP_001158146.1 | 8 | 1044 | ||
NM_001164675.1 | NP_001158147.1 | 8 | 1059 | |||
NM_182760.3 | NP_877437.2 | 9 | 1119 | |||
GenBank mRNA | AB448737.1 | - | 9 | 1090 | ||
AK057983.1 | - | 10 | 1018 | |||
AK075459.1 | - | 9 | 1101 | |||
AK094111.1 | - | 1 | 806 | |||
AK295240.1 | - | 5 | 710 | |||
AK302018.1 | - | 8 | 1012 | |||
AY208752.1 | - | 9 | 1082 | |||
AY323910.1 | - | 9 | 1082 | |||
BC017005.2 | - | 4 | 358 | |||
BC110862.1 | - | 9 | 1090 | 5760490 | ||
BC121122.2 | - | 9 | 1111 | 40120670 | ||
BC121123.2 | - | 8 | 1051 | 40120671 |