Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 645 | ||
NM_173666.3 | NP_775937.1 | 6 | 866 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 2259 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 758 | |||
GenBank mRNA | AK091296.1 | - | 6 | 806 | ||
BC114508.1 | - | 6 | 802 | 40024023 | ||
BC114955.1 | - | 6 | 802 | 40024022 | ||
BX648369.1 | - | 6 | 645 |