Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL28A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037763.2 | NP_001032852.2 | 34 | 3204 | ||
XM_011515358.2 | XP_011513660.1 | 34 | 8049 | |||
XM_011515359.2 | XP_011513661.1 | 34 | 3207 | |||
XM_011515360.2 | XP_011513662.1 | 34 | 4489 | |||
XM_011515362.2 | XP_011513664.1 | 22 | 2009 | |||
XM_017012131.1 | XP_016867620.1 | 35 | 9225 | |||
XM_017012132.1 | XP_016867621.1 | 34 | 4489 | |||
GenBank mRNA | AJ890451.1 | - | 34 | 3204 | ||
BC063866.1 | - | 2 | 138 | |||
BC136892.1 | - | 34 | 3194 | 9020883 | ||
BC144431.1 | - | 35 | 3379 |