Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL28A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037763.2 | NP_001032852.2 | 32 | 2630 | ||
XM_011515358.2 | XP_011513660.1 | 32 | 7475 | |||
XM_011515359.2 | XP_011513661.1 | 32 | 2633 | |||
XM_011515360.2 | XP_011513662.1 | 32 | 3915 | |||
XM_011515362.2 | XP_011513664.1 | 20 | 1435 | |||
XM_017012131.1 | XP_016867620.1 | 33 | 8651 | |||
XM_017012132.1 | XP_016867621.1 | 32 | 3915 | |||
GenBank mRNA | AJ890451.1 | - | 32 | 2630 | ||
BC136892.1 | - | 32 | 2620 | 9020883 | ||
BC144431.1 | - | 33 | 2805 |