Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091367.1 | - | 1 | 58 | ||
BC101445.1 | - | 1 | 58 | |||
BC101446.1 | - | 1 | 58 | |||
BC101447.1 | - | 1 | 58 | |||
BC101985.1 | - | 1 | 58 | |||
BC113049.1 | - | 1 | 58 |