Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006913.3 | NP_008844.1 | 1, 2 | 265 | ||
GenBank mRNA | AB056869.1 | - | 1, 2 | 138 | ||
AK296212.1 | - | 1, 2 | 153 | |||
AK311859.1 | - | 1, 2 | 148 | |||
AL555609.3 | - | 1, 2 | 291 | |||
BC004155.2 | - | 1 | 142 | 2822537 | ||
BC111392.1 | - | 1, 2 | 259 | 6458319 | ||
BC119741.1 | - | 1, 2 | 158 | 40119199 | ||
BC119742.1 | - | 1, 2 | 158 | 40119200 | ||
BC127651.1 | - | 1, 2 | 165 | 40119201 | ||
BC127652.1 | - | 1, 2 | 165 | 40119202 | ||
BC148255.1 | - | 1, 2 | 165 | 40119203 | ||
BT007105.1 | - | 1, 2 | 138 | |||
BU599297.1 | - | 1, 2 | 258 |
Gene Symbol: RNF5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003129.1 | 1 | 195 |