Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096398.1 | - | 1 | 479 | ||
BC104254.1 | - | 1 | 327 | |||
BC104255.1 | - | 1 | 324 | |||
BC113007.1 | - | 1 | 327 | |||
BC113008.1 | - | 1 | 327 |