Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181787.2 | NP_861452.2 | 13, 14 | 1588 | ||
XM_005250894.4 | XP_005250951.2 | 13, 14 | 1801 | |||
XM_005250895.3 | XP_005250952.1 | 12, 13 | 1469 | |||
XM_011516983.2 | XP_011515285.1 | 12, 13 | 1495 | |||
XM_011516984.2 | XP_011515286.1 | 12, 13 | 1468 | |||
XM_017013351.1 | XP_016868840.1 | 12, 13 | 1462 | |||
XM_017013352.1 | XP_016868841.1 | 11, 12 | 1539 | |||
XM_017013353.1 | XP_016868842.1 | 11, 12 | 1363 | |||
XM_017013354.1 | XP_016868843.1 | 12, 13 | 1494 | |||
XM_017013355.1 | XP_016868844.1 | 12, 13 | 1487 | |||
XM_017013356.1 | XP_016868845.1 | 11, 12 | 1340 | |||
XM_017013357.1 | XP_016868846.1 | 11, 12 | 1328 | |||
GenBank mRNA | AK123618.1 | - | 5, 6 | 583 | ||
AK123682.1 | - | 13, 14 | 1542 | |||
AL832859.1 | - | 7, 8 | 789 | |||
BC110870.1 | - | 13, 14 | 1516 | 5580071 | ||
BC126193.1 | - | 13, 14 | 1484 | 8991909 | ||
BC130576.1 | - | 13, 14 | 1484 | 40146607 | ||
BX538048.1 | - | 13, 14 | 1536 | |||
BX538174.1 | - | 13, 14 | 1540 |