Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM133A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171109.1 | NP_001164580.1 | 4 | 2836 | ||
NM_001171110.1 | NP_001164581.1 | 5 | 2871 | |||
NM_001171111.1 | NP_001164582.1 | 3 | 2747 | |||
NM_173698.2 | NP_775969.1 | 4 | 2816 | |||
XM_006724641.3 | XP_006724704.1 | 3 | 2848 | |||
XM_011530925.2 | XP_011529227.1 | 4 | 2789 | |||
XM_011530926.2 | XP_011529228.1 | 4 | 2627 | |||
XM_011530927.2 | XP_011529229.1 | 4 | 2666 | |||
XM_011530928.1 | XP_011529230.1 | 4 | 2600 | |||
XM_011530929.1 | XP_011529231.1 | 5 | 2635 | |||
XM_011530931.2 | XP_011529233.1 | 3 | 2802 | |||
XM_017029454.1 | XP_016884943.1 | 5 | 5475 | |||
XM_017029455.1 | XP_016884944.1 | 6 | 2808 | |||
XM_017029456.1 | XP_016884945.1 | 4 | 2743 | |||
GenBank mRNA | AK094978.1 | - | 4 | 2623 |