Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 3 | 976 | ||
XM_011537432.2 | XP_011535734.1 | 3 | 976 | |||
XM_017021254.1 | XP_016876743.1 | 3 | 976 | |||
XM_017021255.1 | XP_016876744.1 | 3 | 976 | |||
XM_017021257.1 | XP_016876746.1 | 3 | 976 | |||
GenBank mRNA | BC038110.2 | - | 2 | 197 |