Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP19-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181607.1 | NP_853638.1 | 1 | 177 | ||
GenBank mRNA | AB096943.1 | - | 1 | 209 | ||
AJ457067.1 | - | 1 | 173 | |||
BC126284.1 | - | 1 | 238 | 8992000 | ||
BC126286.1 | - | 1 | 238 | 8992002 |