Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204883.1 | NP_001191812.1 | 2 | 85 | ||
NM_001204884.1 | NP_001191813.1 | 2 | 73 | |||
NM_001204885.1 | NP_001191814.1 | 2 | 63 | |||
NM_001204886.1 | NP_001191815.1 | 2 | 100 | |||
NM_001204887.1 | NP_001191816.1 | 1 | 351 | |||
NM_001204888.1 | NP_001191817.1 | 1 | 351 | |||
NM_198490.2 | NP_940892.1 | 1 | 351 | |||
GenBank mRNA | AK128345.1 | - | 1 | 161 | ||
AK291014.1 | - | 1 | 161 | |||
AY166852.2 | - | 1 | 329 | |||
BC062319.1 | - | 1 | 187 | 4873224 | ||
D45418.1 | - | 1 | 140 | |||
DA008447.1 | - | 2 | 73 | |||
DA103180.1 | - | 2 | 63 | |||
DA317502.1 | - | 2 | 65 | |||
DA339537.1 | - | 2 | 100 | |||
DA757569.1 | - | 2 | 85 | |||
DB220260.1 | - | 1 | 352 |