Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207360.2 | NP_997243.2 | 2 | 503 | ||
GenBank mRNA | AB458221.1 | - | 2 | 481 | ||
AB458222.1 | - | 2 | 481 | |||
AK127932.1 | - | 2 | 385 | |||
BC127762.1 | - | 1 | 286 | 40127431 | ||
BC157832.1 | - | 2 | 503 | 9057051 |