Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129820.1 | NP_001123292.1 | 4 | 2026 | ||
XM_017024576.1 | XP_016880065.1 | 6 | 2195 | |||
XM_017024577.1 | XP_016880066.1 | 6 | 2217 | |||
XM_017024578.1 | XP_016880067.1 | 5 | 2084 | |||
XM_017024579.1 | XP_016880068.1 | 5 | 2117 | |||
GenBank mRNA | BC140847.1 | - | 4 | 2023 | 9021715 | |
BC157877.1 | - | 4 | 2027 | 9057094 | ||
BC157879.1 | - | 4 | 2034 | 9057095 |