Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195076.1 | NP_001182005.1 | 3 | 189 | ||
XM_011526910.2 | XP_011525212.1 | 3 | 379 | |||
GenBank mRNA | BX281890.1 | - | 2 | 132 |