Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164688.1 | NP_001158160.1 | 2 | 1395 | ||
NM_183059.2 | NP_898882.1 | 2 | 1398 | |||
XM_017001151.1 | XP_016856640.1 | 1 | 765 | |||
GenBank mRNA | AK291210.1 | - | 2 | 512 | ||
AY191519.1 | - | 2 | 1398 | |||
BC065541.1 | - | 2 | 530 | 6140075 | ||
BX647803.1 | - | 2 | 614 |