Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032290.3 | NP_115666.2 | 4 | 393 | ||
XM_017009978.1 | XP_016865467.1 | 4 | 574 | |||
XM_017009979.1 | XP_016865468.1 | 4 | 5155 | |||
XM_017009980.1 | XP_016865469.1 | 4 | 575 | |||
XM_017009981.1 | XP_016865470.1 | 4 | 425 | |||
XM_017009982.1 | XP_016865471.1 | 4 | 574 | |||
GenBank mRNA | AK297452.1 | - | 4 | 401 | ||
AK299080.1 | - | 4 | 419 | |||
BC054885.1 | - | 4 | 395 | |||
BC070332.1 | - | 4 | 431 | |||
BC107571.1 | - | 4 | 393 | |||
CR936768.1 | - | 4 | 399 | |||
DR001089.1 | - | 4 | 393 |