Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101357.1 | NP_001094827.1 | 2 | 605 | ||
XM_005262409.4 | XP_005262466.1 | 3 | 552 | |||
XM_005262410.4 | XP_005262467.1 | 3 | 473 | |||
GenBank mRNA | BC017958.1 | - | 1 | 856 |