Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145020.1 | NP_001138492.1 | 5 | 546 | ||
NR_026736.1 | 6 | 611 | ||||
NR_026737.1 | 4 | 464 | ||||
XM_006715088.3 | XP_006715151.1 | 6 | 805 | |||
XM_011514572.2 | XP_011512874.1 | 5 | 601 | |||
XM_011514574.2 | XP_011512876.1 | 4 | 511 | |||
XM_011514575.2 | XP_011512877.1 | 6 | 787 | |||
GenBank mRNA | AI376550.1 | - | 1 | 70 | ||
AI621292.1 | - | 1 | 79 | |||
AI809192.1 | - | 1 | 70 | |||
AW183566.1 | - | 1 | 72 | |||
AW293245.1 | - | 1 | 87 | |||
BC016820.1 | - | 4 | 360 |