Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDDC1 SIRNA Assays ›
Gene Symbol:
PDDC1
Gene Name:
Parkinson disease 7 domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 767095 - 777502 on Build GRCh38
UniGene ID:
Hs.218362
Species:
Human
siRNA ID
HSS156312
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318818.1
NP_001305747.1
2
149
NM_001318820.1
NP_001305749.1
3
226
NM_001318821.1
NP_001305750.1
3
226
NM_001318822.1
NP_001305751.1
2
149
NM_001318823.1
NP_001305752.1
2
149
NM_001318824.1
NP_001305753.1
3
226
NM_182612.3
NP_872418.1
3
226
NR_134867.1
2
149
NR_134868.1
3
226
XM_005252898.2
XP_005252955.1
3
211
XM_011520064.1
XP_011518366.1
3
208
XM_017017660.1
XP_016873149.1
3
211
XM_017017661.1
XP_016873150.1
3
208
XM_017017662.1
XP_016873151.1
3
208
XM_017017663.1
XP_016873152.1
3
219
XM_017017664.1
XP_016873153.1
3
211
XM_017017665.1
XP_016873154.1
3
211
XM_017017666.1
XP_016873155.1
2
132
XM_017017667.1
XP_016873156.1
3
225
XM_017017668.1
XP_016873157.1
2
132
GenBank mRNA
AK091602.1
-
3
208
AK092816.1
-
3
211
AK128653.1
-
3
225
AK293559.1
-
2
134
AK309153.1
-
2
132
BC110868.1
-
3
195
5726838
BC112145.1
-
3
207
8327613
BC143415.1
-
2
130
BC143416.1
-
3
207
9051924
BC143418.1
-
3
207
9051926
BC143419.1
-
2
130
BC143420.1
-
3
207
9051928
BC143421.1
-
3
207
BC143422.1
-
2
130
BC143423.1
-
2
130
DA025348.1
-
3
211
DC364167.1
-
3
226
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
lactate biosynthetic process
molecular_function
glyoxalase III activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline