Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDDC1 SIRNA Assays ›
Gene Symbol:
PDDC1
Gene Name:
Parkinson disease 7 domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 767095 - 777502 on Build GRCh38
UniGene ID:
Hs.218362
Species:
Human
siRNA ID
HSS156314
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318818.1
NP_001305747.1
5
470
NM_001318820.1
NP_001305749.1
5
439
NM_001318821.1
NP_001305750.1
6
547
NM_001318822.1
NP_001305751.1
4
362
NM_001318823.1
NP_001305752.1
5
470
NM_001318824.1
NP_001305753.1
5
439
NM_182612.3
NP_872418.1
6
547
NR_134867.1
5
470
NR_134868.1
6
641
XM_005252898.2
XP_005252955.1
6
573
XM_011520064.1
XP_011518366.1
6
529
XM_017017660.1
XP_016873149.1
6
532
XM_017017661.1
XP_016873150.1
6
529
XM_017017663.1
XP_016873152.1
5
432
XM_017017664.1
XP_016873153.1
6
573
XM_017017665.1
XP_016873154.1
6
573
XM_017017666.1
XP_016873155.1
5
453
XM_017017667.1
XP_016873156.1
5
479
XM_017017668.1
XP_016873157.1
5
453
GenBank mRNA
AK091602.1
-
6
529
AK092816.1
-
5
424
AK128653.1
-
5
479
AK293559.1
-
5
455
AK309153.1
-
5
453
BC110868.1
-
6
516
5726838
BC112145.1
-
6
528
8327613
BC143415.1
-
4
343
BC143416.1
-
5
420
9051924
BC143418.1
-
6
528
9051926
BC143419.1
-
5
451
BC143420.1
-
5
420
9051928
BC143421.1
-
6
622
BC143422.1
-
5
451
BC143423.1
-
5
451
DA025348.1
-
6
532
DC364167.1
-
6
547
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
lactate biosynthetic process
molecular_function
glyoxalase III activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0