Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012759.2 | NP_001012777.1 | 6 | 436 | ||
NM_001012762.2 | NP_001012780.1 | 6 | 436 | |||
NM_001318507.1 | NP_001305436.1 | 6 | 649 | |||
NM_001318513.1 | NP_001305442.1 | 5 | 357 | |||
XM_017023210.1 | XP_016878699.1 | 1 | 16 | |||
GenBank mRNA | AK299521.1 | - | 396 | |||
BC021056.1 | - | 6 | 380 | |||
BC080540.1 | - | 6 | 344 | |||
BC108659.1 | - | 6 | 384 | 6167465 | ||
BC121805.1 | - | 5 | 337 | 40107956 | ||
BC125269.1 | - | 6 | 416 | 40107964 | ||
BC157881.1 | - | 6 | 397 | 8862703 | ||
BI520274.1 | - | 6 | 436 | |||
BQ430809.1 | - | 416 |