Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012759.2 | NP_001012777.1 | 7 | 767 | ||
NM_001012762.2 | NP_001012780.1 | 7 | 767 | |||
NM_001318507.1 | NP_001305436.1 | 7 | 980 | |||
NM_001318513.1 | NP_001305442.1 | 6 | 688 | |||
XM_017023210.1 | XP_016878699.1 | 2 | 325 | |||
GenBank mRNA | BC021056.1 | - | 7 | 711 | ||
BC080540.1 | - | 7 | 675 | |||
BC108659.1 | - | 7 | 715 | 6167465 | ||
BC121805.1 | - | 6 | 668 | 40107956 | ||
BC125269.1 | - | 7 | 747 | 40107964 | ||
BC157881.1 | - | 7 | 728 | 8862703 |
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL582912.3 | - | 9 | 935 |