Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021250.3 | NP_067073.1 | 5 | 650 | ||
NM_181879.2 | NP_870994.1 | 5 | 650 | |||
NM_181985.3 | NP_871714.1 | 4 | 614 | |||
NM_181986.2 | NP_871715.1 | 4 | 614 | |||
GenBank mRNA | AF212842.1 | - | 2 | 364 | ||
AF324830.1 | - | 5 | 650 | |||
AF499916.1 | - | 5 | 593 | |||
AF499917.1 | - | 4 | 549 | |||
AF499918.1 | - | 5 | 609 | |||
AF499919.1 | - | 4 | 566 | |||
EU915611.1 | - | 5 | 590 | |||
EU915612.1 | - | 4 | 554 |