Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002208.1 | 1 | 362 | |||
GenBank mRNA | BM979118.1 | - | 1 | 321 |
Gene Symbol: C15orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289132.1 | NP_001276061.1 | 4 | 794 | ||
GenBank mRNA | AK098781.1 | - | 4 | 729 |