Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202439.2 | NP_001189368.1 | 4 | 1003 | ||
XM_011520072.2 | XP_011518374.1 | 4 | 2957 | |||
XM_011520073.2 | XP_011518375.1 | 4 | 2703 | |||
XM_011520074.2 | XP_011518376.1 | 4 | 1320 | |||
XM_011520075.2 | XP_011518377.1 | 4 | 2609 | |||
GenBank mRNA | BC136797.1 | - | 4 | 866 | ||
BC136800.1 | - | 4 | 866 | |||
BX537685.1 | - | 4 | 995 | |||
CR749521.1 | - | 4 | 1003 |