Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256508.1 | NP_001243437.1 | 7 | 654 | ||
NM_198517.3 | NP_940919.1 | 7 | 675 | |||
NR_046266.1 | 6 | 845 | ||||
XM_006718538.3 | XP_006718601.1 | 7 | 934 | |||
XM_006718539.3 | XP_006718602.1 | 7 | 917 | |||
XM_006718540.3 | XP_006718603.1 | 7 | 847 | |||
XM_006718541.3 | XP_006718604.1 | 7 | 938 | |||
XM_006718542.3 | XP_006718605.1 | 7 | 921 | |||
XM_006718543.2 | XP_006718606.1 | 7 | 655 | |||
XM_006718544.3 | XP_006718607.1 | 7 | 849 | |||
XM_011545001.2 | XP_011543303.1 | 6 | 876 | |||
XM_011545002.2 | XP_011543304.1 | 6 | 895 | |||
GenBank mRNA | AK122585.1 | - | 2 | 1327 | ||
BC022202.1 | - | 6 | 619 | |||
BC036873.1 | - | 6 | 630 | 5224063 | ||
BC062999.1 | - | 7 | 647 | 5176854 |