Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199046.2 | NP_950247.2 | 2, 3 | 415 | ||
NM_199456.2 | NP_955535.2 | 2, 3 | 415 | |||
XM_017023218.1 | XP_016878707.1 | 2, 3 | 271 | |||
GenBank mRNA | AY363222.1 | - | 2, 3 | 415 | ||
AY363223.1 | - | 2, 3 | 415 | |||
BC104458.1 | - | 2, 3 | 406 | 40030655 | ||
BC104459.1 | - | 2, 3 | 406 | 40030657 |