Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145416.1 | NP_001138888.1 | 2 | 212 | ||
NM_001321112.1 | NP_001308041.1 | 2 | 321 | |||
NM_001321113.1 | NP_001308042.1 | 2 | 131 | |||
NM_001321114.1 | NP_001308043.1 | 2 | 240 | |||
NM_033408.3 | NP_212133.1 | 2 | 331 | |||
NM_198536.2 | NP_940938.1 | 1 | 710 | |||
XM_005259899.2 | XP_005259956.1 | 2 | 417 | |||
GenBank mRNA | AK127147.1 | - | 2 | 2493 | ||
AK291979.1 | - | 2 | 191 | |||
AY358949.1 | - | 2 | 152 | |||
BC064948.1 | - | 1 | 451 | 6025162 | ||
BC091472.1 | - | 2 | 139 | 30335220 | ||
BE889078.1 | - | 2 | 331 | |||
BM352837.1 | - | 2 | 90 | |||
BM451378.1 | - | 2 | 217 | |||
BQ639667.1 | - | 2 | 248 | |||
CB994930.1 | - | 2 | 166 |