Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080471.1 | NP_001073940.1 | 11, 12 | 1529 | ||
XM_005245141.3 | XP_005245198.1 | 12, 13 | 1722 | |||
XM_011509510.2 | XP_011507812.1 | 12, 13 | 1661 | |||
XM_011509511.2 | XP_011507813.1 | 11, 12 | 1617 | |||
XM_011509512.2 | XP_011507814.1 | 11, 12 | 1541 | |||
XM_017001234.1 | XP_016856723.1 | 11, 12 | 2787 | |||
XM_017001235.1 | XP_016856724.1 | 11, 12 | 2657 | |||
XM_017001236.1 | XP_016856725.1 | 11, 12 | 1624 | |||
XM_017001237.1 | XP_016856726.1 | 12, 13 | 1868 | |||
XM_017001238.1 | XP_016856727.1 | 12, 13 | 1807 | |||
XM_017001239.1 | XP_016856728.1 | 12, 13 | 1618 | |||
XM_017001240.1 | XP_016856729.1 | 9, 10 | 1186 | |||
XM_017001241.1 | XP_016856730.1 | 9, 10 | 1334 | |||
XM_017001242.1 | XP_016856731.1 | 9, 10 | 1238 | |||
GenBank mRNA | AK074121.1 | - | 6, 7 | 1142 |