Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014336.1 | NP_001014358.1 | 3 | 508 | ||
GenBank mRNA | AY499343.1 | - | 3 | 508 | ||
BC132998.1 | - | 3 | 502 | 40147020 | ||
BC133000.1 | - | 3 | 502 | 40147022 |