Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300935.1 | NP_001287864.1 | 3 | 481 | ||
NM_207172.1 | NP_997055.1 | 3 | 435 | |||
NM_207173.1 | NP_997056.1 | 3 | 435 | |||
GenBank mRNA | AY310326.1 | - | 3 | 435 | ||
AY310327.1 | - | 3 | 435 | |||
AY310331.1 | - | 3 | 435 | |||
AY956448.1 | - | 3 | 311 | |||
BC132693.1 | - | 3 | 352 | 40146715 | ||
BC132695.1 | - | 3 | 352 | 40146717 |
Set Membership: |
G-protein coupled receptor |