Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002594.1 | 1 | 402 | |||
GenBank mRNA | BC071656.1 | - | 1 | 375 | ||
BI758996.1 | - | 1 | 149 | |||
BM904244.1 | - | 374 |
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135317.1 | 1 | 400 | |||
GenBank mRNA | AB055226.1 | - | 1 | 402 |