Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130934.1 | 2, 3 | 256 | |||
GenBank mRNA | AB111913.1 | - | 2, 3 | 256 | ||
BC040611.2 | - | 2, 3 | 180 | |||
BC130491.1 | - | 2, 3 | 180 | |||
BC130493.1 | - | 2, 3 | 180 |