Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010923.2 | NP_001010923.1 | 1 | 203 | ||
NM_001164685.1 | NP_001158157.1 | 1 | 203 | |||
XM_011535813.2 | XP_011534115.1 | 2 | 370 | |||
XM_011535816.1 | XP_011534118.1 | 1 | 170 | |||
XM_017010848.1 | XP_016866337.1 | 2 | 370 | |||
XM_017010849.1 | XP_016866338.1 | 2 | 370 | |||
GenBank mRNA | AK097903.1 | - | 1 | 180 | ||
AK292046.1 | - | 1 | 153 | |||
BC130516.1 | - | 1 | 197 | 40146547 | ||
BC144034.1 | - | 1 | 195 | |||
BX956657.1 | - | 2 | 407 | |||
DA818250.1 | - | 2 | 371 |