Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162505.1 | NP_001155977.1 | 5, 6 | 842 | ||
NM_199129.2 | NP_954580.1 | 5, 6 | 851 | |||
NR_027889.1 | 6, 7 | 955 | ||||
GenBank mRNA | AB044550.1 | - | 5, 6 | 764 | ||
AF155120.1 | - | 5, 6 | 778 | |||
AK023028.1 | - | 5, 6 | 776 | |||
AK304595.1 | - | 4, 5 | 566 | |||
BC018893.2 | - | 5, 6 | 763 | |||
BC142966.1 | - | 5, 6 | 766 | 8860386 | ||
BC144612.1 | - | 6, 7 | 955 |
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF982644.1 | - | 4, 5 | 498 | ||
BI838424.1 | - | 5, 6 | 767 |