Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 12 | 4059 | |||
GenBank mRNA | AB209623.1 | - | 12 | 3847 |
Gene Symbol: C1orf158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647383.1 | - | 1579 |
Gene Symbol: DISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522125.2 | XP_011520427.1 | 10 | 9639 | ||
XM_017022687.1 | XP_016878176.1 | 11 | 9388 |
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004818.1 | - | 18 | 2672 |
Gene Symbol: LIX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153713.2 | NP_714924.1 | 6 | 2323 | ||
XM_017000316.1 | XP_016855805.1 | 5 | 2246 | |||
GenBank mRNA | AK123675.1 | - | 3 | 1575 | ||
AK128733.1 | - | 7 | 2164 | |||
CR627381.1 | - | 5 | 1962 |
Gene Symbol: PKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118089.2 | - | 11 | 2280 |