Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144869.1 | NP_001138341.1 | 1 | 442 | ||
XM_011545021.2 | XP_011543323.2 | 1 | 704 | |||
GenBank mRNA | CN348282.1 | - | 1 | 448 | ||
CX752266.1 | - | 1 | 424 |