Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005353.2 | NP_001005353.1 | 5 | 858 | ||
NM_013410.3 | NP_037542.1 | 4 | 741 | |||
NM_203464.2 | NP_982289.1 | 5 | 747 | |||
XM_017000612.1 | XP_016856101.1 | 4 | 493 | |||
XM_017000613.1 | XP_016856102.1 | 4 | 679 | |||
GenBank mRNA | AK025926.1 | - | 4 | 705 | ||
AK309057.1 | - | 4 | 449 | |||
AK313611.1 | - | 4 | 705 | |||
BC016180.1 | - | 4 | 641 | 4637988 | ||
BC040224.1 | - | 4 | 617 | 5188877 | ||
BC066944.1 | - | 5 | 858 | 4819382 | ||
BC136886.1 | - | 4 | 582 | 9020877 | ||
BC136887.1 | - | 1 | 582 | 9020878 | ||
BC146653.1 | - | 5 | 732 | 40147793 | ||
BC148270.1 | - | 4 | 522 | 8860213 | ||
CR456830.1 | - | 4 | 500 | |||
X60673.1 | - | 4 | 683 |