Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172623.1 | NP_001166094.1 | 28 | 6718 | ||
NM_001172624.1 | NP_001166095.1 | 28 | 6844 | |||
NM_002499.3 | NP_002490.2 | 29 | 6877 | |||
XM_005254408.1 | XP_005254465.1 | 30 | 7187 | |||
XM_011521628.2 | XP_011519930.1 | 29 | 6942 | |||
XM_011521629.2 | XP_011519931.1 | 28 | 6903 | |||
XM_011521630.2 | XP_011519932.1 | 29 | 6888 | |||
XM_011521632.2 | XP_011519934.1 | 27 | 6834 | |||
XM_011521633.2 | XP_011519935.1 | 28 | 6777 | |||
XM_017022231.1 | XP_016877720.1 | 28 | 6795 | |||
XM_017022232.1 | XP_016877721.1 | 29 | 7106 | |||
XM_017022233.1 | XP_016877722.1 | 27 | 6744 | |||
XM_017022234.1 | XP_016877723.1 | 29 | 6772 | |||
XM_017022235.1 | XP_016877724.1 | 27 | 6696 | |||
XM_017022236.1 | XP_016877725.1 | 27 | 6684 | |||
XM_017022237.1 | XP_016877726.1 | 26 | 6675 | |||
XM_017022238.1 | XP_016877727.1 | 27 | 6636 | |||
GenBank mRNA | AB209412.1 | - | 24 | 5692 | ||
AL355708.1 | - | 1 | 1485 | |||
AL359597.1 | - | 1 | 1047 | |||
AL390182.1 | - | 1 | 1485 |