Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 4792 | ||
XM_006720544.3 | XP_006720607.1 | 16 | 4768 | |||
XM_017022240.1 | XP_016877729.1 | 18 | 4663 | |||
XM_017022241.1 | XP_016877730.1 | 17 | 4148 | |||
XM_017022243.1 | XP_016877732.1 | 17 | 4144 | |||
XM_017022244.1 | XP_016877733.1 | 17 | 4046 | |||
XM_017022245.1 | XP_016877734.1 | 18 | 4101 | |||
XM_017022246.1 | XP_016877735.1 | 17 | 4202 | |||
XM_017022251.1 | XP_016877740.1 | 10 | 3163 | |||
XM_017022252.1 | XP_016877741.1 | 9 | 3139 | |||
XM_017022253.1 | XP_016877742.1 | 18 | 4834 | |||
XM_017022254.1 | XP_016877743.1 | 18 | 4867 |