Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 2 | 518 | ||
NM_001077418.2 | NP_001070886.1 | 3 | 421 | |||
NR_074083.1 | 3 | 544 | ||||
GenBank mRNA | AK025820.1 | - | 3 | 393 | ||
AK096650.1 | - | 2 | 470 | |||
AK290483.1 | - | 3 | 407 | |||
AK307263.1 | - | 3 | 510 | |||
AY358612.1 | - | 3 | 417 | |||
BC010609.1 | - | 3 | 375 | 4214587 | ||
BC016401.1 | - | 2 | 53 | |||
BC063677.1 | - | 3 | 526 | |||
BU173659.1 | - | 3 | 485 | |||
DC330418.1 | - | 3 | 421 |