Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 2 | 589 | ||
NM_001077418.2 | NP_001070886.1 | 3 | 492 | |||
NR_074083.1 | 3 | 615 | ||||
GenBank mRNA | AK025820.1 | - | 3 | 464 | ||
AK096650.1 | - | 2 | 541 | |||
AK290483.1 | - | 3 | 478 | |||
AK307263.1 | - | 3 | 581 | |||
AY358612.1 | - | 3 | 488 | |||
BC010609.1 | - | 3 | 446 | 4214587 | ||
BC016401.1 | - | 2 | 124 | |||
BC063677.1 | - | 3 | 597 | |||
BU173659.1 | - | 3 | 556 | |||
DC330418.1 | - | 3 | 492 |