Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013641.2 | NP_001013663.1 | 6 | 1029 | ||
GenBank mRNA | BC127706.1 | - | 6 | 1009 | 40127826 | |
BC127707.1 | - | 6 | 1012 | 40127833 | ||
BC137239.1 | - | 6 | 1029 | 9021236 | ||
BC137240.1 | - | 6 | 1029 | 9021237 |